Ethyl-1-bromocyclobutanecarboxylate 96%

Supplier: Thermo Fisher Scientific
Warning

188710010 188710050
ACRO188710010EA 24.3 EUR
ACRO188710010 ACRO188710050
Ethyl-1-bromocyclobutanecarboxylate 96%
Ethyl-1-bromocyclobutanecarboxylate

Appearance: Clear colorless Liquid

Formula: C₇H₁₁BrO₂
Boiling Pt: 85…88 °C (12 mmHg)
Density: 1,279 g/cm³
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001322
CAS Number: 35120-18-4

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4700 to 1.4720 (20°C, 589 nm)
GC ≥95 %
HPLC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR