1-Ethylpropylamine 98+%

Supplier: Thermo Fisher Scientific
Danger

183700050 183700250 183701000
ACRO183700050EA 36.7 EUR
ACRO183700050 ACRO183700250 ACRO183701000
1-Ethylpropylamine 98+%
1-Ethylpropylamine
Formula: C₅H₁₃N
Boiling Pt: 90 °C (1013 hPa)
Density: 0,74 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00008096
CAS Number: 616-24-0
UN: 1106
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4040 to 1.4060 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR