4-(Chloromethyl)benzonitrile 98%

Supplier: Thermo Fisher Scientific
Danger

391150100 391150500
ACRO391150100EA 65 EUR
ACRO391150100 ACRO391150500
4-(Chloromethyl)benzonitrile 98%
4-(Chloromethyl)benzonitrile
Formula: C₈H₆ClN
MW: 151.60 g/mol
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00019761
CAS Number: 874-86-2
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 77 to 80 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR