Bromomethyl acetate 95%

Supplier: Thermo Fisher Scientific
Warning

399180010 399180100
ACRO399180010EA 124 EUR
ACRO399180010 ACRO399180100
Bromomethyl acetate 95%
Bromomethyl acetate
Formula: H₃CCOOCH₂Br
Boiling Pt: 130…133 °C (750 mmHg)
Flash Pt: 57 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000170
CAS Number: 590-97-6
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4470 to 1.4540 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR