[Hydroxy(tosyloxy)iodo]benzene 97+%

Supplier: Thermo Fisher Scientific
Warning

389610050 389610250
ACRO389610050EA 50.9 EUR
ACRO389610050 ACRO389610250
[Hydroxy(tosyloxy)iodo]benzene 97+%
[Hydroxy(tosyloxy)iodo]benzene

Appearance: Light yellow Crystalline powder

Formula: C₁₃H₁₃IO₄S
Melting Pt: 131…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00011547
CAS Number: 27126-76-7

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR