Allyl methyl carbonate 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Carbonic acid Allyl Methyl Ester, Allyl Methyl Carbonate
389750050
389750250
ACRO389750050EA
94.8
EUR
ACRO389750050
ACRO389750250
Allyl methyl carbonate 98%
Allyl methyl carbonate
Formula:
C₅H₈O₃ MW: 116,1152 g/mol Flash Pt: 33 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134545 CAS Number: 35466-83-2 UN: 3272 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to almost colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4050 to 1.4090 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...