(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid 98%

Supplier: Thermo Scientific

Synonyms: N-Boc-2-cyclohexyl-L-glycine, Boc-cyclohexyl-Gly-OH, BOC-L-α-cyclohexylglycine, N-Boc-L-2-cyclohexylglycine, Boc-Chg-OH

399350010 399350050
ACRO399350010EA 59.4 EUR
ACRO399350010 ACRO399350050
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid 98%
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid

Appearance: White Powder

Formula: C₁₃H₂₃NO₄
MW: 257,3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00133638
CAS Number: 109183-71-3

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
NMR consistent with structure
Specific optical rotation +8° to +11° (20°C, 589 nm) (c=1,CH3OH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR