Benzylphosphonic acid 98%
Supplier: Thermo Scientific
|
Danger
|
393340010
393340050
393340250
ACRO393340010EA
39.7
EUR
ACRO393340010
ACRO393340050
ACRO393340250
Benzylphosphonic acid 98%
Benzylphosphonic acid
Formula:
C₇H₉O₃P Melting Pt: 165…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039519 CAS Number: 6881-57-8 UN: 3261 ADR: 8,III |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Melting point | 169 to 173 °C |
Titration with NaOH | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...