4-Methoxy-2-methylbenzaldehyde 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-Methyl-4-methoxybenzaldehyde
427020010
427020050
ACRO427020010EA
52.7
EUR
ACRO427020010
ACRO427020050
4-Methoxy-2-methylbenzaldehyde 97%
4-Methoxy-2-methylbenzaldehyde
Formula:
C₉H₁₀O₂ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD02261771 CAS Number: 52289-54-0 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...