DL-Pantolactone 97%

Supplier: Thermo Fisher Scientific
Warning

427590250 427591000
ACRO427590250EA 59.3 EUR
ACRO427590250 ACRO427591000
DL-Pantolactone 97%
DL-Pantolactone

Appearance: White to light yellow Powder

Formula: C₆H₁₀O₃
Boiling Pt: 246 °C (760 mmHg)
Melting Pt: 74…78 °C
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00064333
CAS Number: 79-50-5

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤0.5 %
GC ≥96.0 %
UV at 240 nm A: ≤0.05
Titration after Ion exchange ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR