2-Formyl-3-thiopheneboronic acid 97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Formylthiophene-3-boronic acid, (2-Formylthiophen-3-yl)boronic acid

427690010 427690050
ACRO427690010EA 44.5 EUR
ACRO427690010 ACRO427690050
2-Formyl-3-thiopheneboronic acid 97%
2-Formyl-3-thiopheneboronic acid
Formula: C₅H₅BO₃S
MW: 155,97 g/mol
Melting Pt: 137…193 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01075678
CAS Number: 4347-31-3

Order Now

Specification Test Results

Appearance White to light beige to light brown Powder or crystals
Infrared spectrum Conforms
HPLC ≥96.0 %
Sulfur (S) ≤0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR