Cutout rings with bosshead
Supplier: R-L ENTERPRISES LTD
2015/2 2015/6 2015/11 2015/17
241-0167EA 7.2 EUR
241-0167 241-0168 241-0165 241-0166
Cutout rings with bosshead
Retort Rings
Mild steel, rubber coated.
  • Standard stem length with various ring diameters
  • Stem length to the centre of the bosshead is approximately 60 mm
  • With welded joints
  • Cut out section allows easy removal of equipment
  • Fitted with a bosshead with a blue plastic headed nickel plated steel screw