(±)-α-Bromophenylacetic acid 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
α-Bromophenylacetic Acid, DL-α-Bromophenylacetic acid
107270050
107270250
107271000
ACRO107270050EA
37.5
EUR
ACRO107270050
ACRO107270250
ACRO107271000
(±)-α-Bromophenylacetic acid 97%
(±)-α-Bromophenylacetic acid
Formula:
C₈H₇BrO₂ MW: 215,0440 g/mol Melting Pt: 82…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004206 CAS Number: 4870-65-9 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...