4,4'-Diiodobiphenyl 99%

Supplier: Thermo Fisher Scientific
Warning

115120050 115120250 115121000
ACRO115120050EA 29.8 EUR
ACRO115120050 ACRO115120250 ACRO115121000
4,4'-Diiodobiphenyl 99%
4,4'-Diiodobiphenyl
Formula: C₁₂H₈I₂
Melting Pt: 201…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00001057
CAS Number: 3001-15-8
UN: 3152
ADR: 9,II

Order Now

Specification Test Results

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Water ≤0.3 % (K.F.)
GC ≥99.0 %
Color scale ≤50 APHA
Acidity ≤0.2 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR