trans-4-Hydroxystilbene 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4-[(E)-2-phenylvinyl]phenol, 4-Styrylphenol, p-HYDROXYSTILBENE
121930050
121930250
ACRO121930050EA
74.8
EUR
ACRO121930050
ACRO121930250
trans-4-Hydroxystilbene 98%
trans-4-Hydroxystilbene
Formula:
C₁₄H₁₂O MW: 196.249 g/mol Melting Pt: 185…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002386 CAS Number: 6554-98-9 |
Specification Test Results
Appearance | Yellow to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 185 to 189 °C |
HPLC | ≥96.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...