2-Chloro-6-nitrobenzaldehyde 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
147750010
147750050
ACRO147750010EA
67.5
EUR
ACRO147750010
ACRO147750050
2-Chloro-6-nitrobenzaldehyde 98%
2-Chloro-6-nitrobenzaldehyde
Formula:
C₇H₄ClNO₃ MW: 185.57 g/mol Melting Pt: 69…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007204 CAS Number: 6361-22-4 |
Specification Test Results
Appearance | White to yellow to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 68 to 73 °C |
GC | ≥97.5 % |
NMR | consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...