5-Bromoisatin 98%

Supplier: Thermo Scientific
Danger

Synonyms: 5-Bromoindoline-2,3-dione, 5-Bromo-1H-indole-2,3-dione

154230050 154230250
ACRO154230050EA 34.2 EUR
ACRO154230050 ACRO154230250
5-Bromoisatin 98%
5-Bromoisatin
Formula: C₈H₄BrNO₂
MW: 226,03 g/mol
Melting Pt: 251…253 °C
Storage Temperature: Ambient
MDL Number: MFCD00149345
CAS Number: 87-48-9

Order Now

Specification Test Results

Appearance Orange Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
GC ≥96 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR