Neopentylamine 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1-Amino-2,2-dimethylpropane

173690250 173691000
ACRO173690250EA 144 EUR
ACRO173690250 ACRO173691000
Neopentylamine 97%
Neopentylamine
Formula: C₅H₁₃N
MW: 87.16 g/mol
Boiling Pt: 79…82 °C (760 mmHg)
Density: 0.74 g/cm³
Flash Pt: –13 °C
Storage Temperature: Ambient
MDL Number: MFCD00008134
CAS Number: 5813-64-9
UN: 2733
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4010 to 1.4050 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR