exo-2-Aminonorbornane 99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(1R,2R,4S)-Bicyclo[2.2.1]heptan-2-amine, (1R,2R,4S)-rel-Bicyclo[2.2.1]heptan-2-amine, (1alpha,4alpha)-Norbornane-2alpha-amine
176950010
176950050
ACRO176950010EA
78.7
EUR
ACRO176950010
ACRO176950050
exo-2-Aminonorbornane 99%
exo-2-Aminonorbornane
Formula:
C₇H₁₃N MW: 111,19 g/mol Boiling Pt: 49 °C (13mbar) Density: 0,93 g/cm³ Flash Pt: 35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00078132 CAS Number: 7242-92-4 UN: 2733 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥98.5 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...