1-Pentanesulphonic acid sodium salt 98+%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Sodium 1-pentanesulphonate, Sodium pentane-1-sulphonate, Pentane-1-sulphonic acid sodium salt, Sodium pentanesulphonate, 98%, Sodium pentanesulphonate
206110050
206110250
206111000
ACRO206110050EA
47.5
EUR
ACRO206110050
ACRO206110250
ACRO206111000
1-Pentanesulphonic acid sodium salt 98+%
1-Pentanesulphonic acid sodium salt
Formula:
C₅H₁₁O₃SNa MW: 174,2 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007541 CAS Number: 22767-49-3 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % (1 g, 120°C) |
Solubility | (10 % in water at 25°C) Clear colorless |
Titration after Ion exchange | ≥98.0 % (on dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...