Nitrofurantoin 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
1-[(5-Nitrofurfurylidene)amino]hydantoin
229380100
229380250
229381000
ACRO229380100EA
35.2
EUR
ACRO229380100
ACRO229380250
ACRO229381000
Nitrofurantoin 98%
Nitrofurantoin
Formula:
C₈H₆N₄O₅ MW: 238.16 g/mol Melting Pt: 268 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003224 CAS Number: 67-20-9 |
Specification Test Results
Appearance | Yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...