Methyl-6-methylnicotinate 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Methyl 6-methylnicotinate
248210020
248210100
ACRO248210020EA
61.4
EUR
ACRO248210020
ACRO248210100
Methyl-6-methylnicotinate 97%
Methyl 6-methylnicotinate
Formula:
C₈H₉NO₂ MW: 151.16 g/mol Boiling Pt: 108…110 °C (22.5 mmHg) Melting Pt: 31…37 °C Flash Pt: 103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006340 CAS Number: 5470-70-2 |
Specification Test Results
Appearance | Brown Low melting solid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...