Pentafluoroiodobenzene 99% Stabilised

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Iodopentafluorobenzene

Stabilisation: Copper

312200010 312200050
ACRO312200010EA 14.7 EUR
ACRO312200010 ACRO312200050
Pentafluoroiodobenzene 99% Stabilised
Pentafluoroiodobenzene
Formula: C₆F₅I
MW: 293.96 g/mol
Boiling Pt: 162…165 °C (760 mmHg)
Melting Pt: -29
Density: 2.204 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00001032
CAS Number: 827-15-6

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR