(3-Cyanopropyl)dimethylchlorosilane 95%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Chloro(3-cyanopropyl)dimethylsilane
313380100
313380500
ACRO313380100EA
78.3
EUR
ACRO313380100
ACRO313380500
(3-Cyanopropyl)dimethylchlorosilane 95%
(3-Cyanopropyl)dimethylchlorosilane
Formula:
C₆H₁₂ClNSi MW: 161,70 g/mol Boiling Pt: 230…235 °C (760 mmHg) Density: 0,99 g/cm³ Flash Pt: 85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001969 CAS Number: 18156-15-5 UN: 2987 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to straw yellow to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4450 to 1.4470 (20°C, 589 nm) |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...