4-Amino-3,5-dichloropyridine 97%

Supplier: Thermo Scientific
Warning

Synonyms: 3,5-Dichloro-4-pyridylamine, 3,5-Dichloropyridin-4-amine

367220050 367220250 367221000
ACRO367220050EA 30.9 EUR
ACRO367220050 ACRO367220250 ACRO367221000
4-Amino-3,5-dichloropyridine 97%
4-Amino-3,5-dichloropyridine
Formula: C₅H₄Cl₂N₂
MW: 163,01 g/mol
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00125023
CAS Number: 22889-78-7

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 156 to 164 °C
Water ≤1 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR