(R)-(-)-2-(Methoxymethyl)pyrrolidine 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(R)-2-(Methoxymethyl)pyrrolidine
376190010
376190050
ACRO376190010EA
105
EUR
ACRO376190010
ACRO376190050
(R)-(-)-2-(Methoxymethyl)pyrrolidine 98%
(R)-(-)-2-(Methoxymethyl)pyrrolidine
Formula:
C₆H₁₃NO Boiling Pt: 62 °C (40 mmHg) Density: 0,932 g/cm³ Flash Pt: 45 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00066219 CAS Number: 84025-81-0 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Specific optical rotation | -32.5° to -36.5° (20°C, 589 nm) (c=2 in aq HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...