Ethyl myristate 98+%
Supplier: Thermo Fisher Scientific
Warning
|
Synonyms:
Tetradecanoic acid ethyl ester, Tetradecylic acid ethyl ester, Ethyl tetradecanoate, Myristic acid ethyl ester
385540250
385541000
385545000
ACRO385540250EA
26.9
EUR
ACRO385540250
ACRO385541000
ACRO385545000
Ethyl myristate 98+%
Ethyl myristate
Formula:
C₁₆H₃₂O₂ MW: 256.43 g/mol Boiling Pt: 295 °C (760 mmHg) Melting Pt: 11…12 °C Density: 0.861 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008984 CAS Number: 124-06-1 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...