2-Hydroxythiophenol 98+%

Supplier: Thermo Fisher Scientific
Danger

385600010 385600050
ACRO385600010EA 82.7 EUR
ACRO385600010 ACRO385600050
2-Hydroxythiophenol 98+%
2-Hydroxythiophenol
Formula: C₆H₆OS
Boiling Pt: 40 °C (300 mmHg)
Melting Pt: 20 °C
Density: 1,22 g/cm³
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00040447
CAS Number: 1121-24-0
UN: 2927
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear yellow to light green Liquid
Infrared spectrum Conforms
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR