2-Hydroxyphenylboronic acid 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Hydroxybenzeneboronic acid, Dihydroxy(2-hydroxyphenyl)borane, (2-Hydroxyphenyl)metaboric acid
387500010
387500050
ACRO387500010EA
76.4
EUR
ACRO387500010
ACRO387500050
2-Hydroxyphenylboronic acid 97%
2-Hydroxyphenylboronic acid
Formula:
C₆H₇BO₃ MW: 137,93 g/mol Melting Pt: 173…174 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01074581 CAS Number: 89466-08-0 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...