Benzyltrimethylammonium tribromide 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Mono(N,N,N-trimethyl-1-phenylmethanaminium) tribromide, Benzyl trimethyl ammonium tribromide
388320100
388320500
ACRO388320100EA
65.5
EUR
ACRO388320100
ACRO388320500
Benzyltrimethylammonium tribromide 97%
Benzyltrimethylammonium tribromide
Formula:
C₁₀H₁₆Br₃N MW: 389,97 g/mol Melting Pt: 97…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134423 CAS Number: 111865-47-5 |
Specification Test Results
Appearance | Yellow to orange Crystalline powder |
Infrared spectrum | Conforms |
Titration Iodometric | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...