Trilithium orthophosphate, extra pure
Supplier: Thermo Fisher Scientific
|
Warning
|
389781000
389785000
ACRO389781000EA
80.7
EUR
ACRO389781000
ACRO389785000
Trilithium orthophosphate, extra pure
tri-Lithium orthophosphate
Formula:
Li₃O₄P Melting Pt: 837 °C Density: 2,41 g/cm³ (15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016187 CAS Number: 10377-52-3 |
Specification Test Results
Appearance | White Crystalline powder |
Titration with HClO4 | ≥95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...