(S)-2-Methylpropane-2-sulphinamide 97+%, >98.0% ee

Supplier: Thermo Fisher Scientific

Synonyms: (S)-tert-Butylsulphinamide, (S)-(-)-tert-Butylsulphinamide, (S)-(-)-t-Butylsulphinamide, (S)-(-)-2-Methyl-2-propanesulphinamide, (S)-(-)-tert-Butyl sulphinamide

391080010 391080050
ACRO391080010EA 85.2 EUR
ACRO391080010 ACRO391080050
(S)-2-Methylpropane-2-sulphinamide 97+%, >98.0% ee
(S)-2-Methylpropane-2-sulphinamide
Formula: C₄H₁₁NOS
Melting Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05861480
CAS Number: 343338-28-3

Order Now

Specification Test Results

Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
GC ≥97.0 %
Enantiomeric excess ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR