4-Iodobiphenyl 97+%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Iodo-1,1'-biphenyl

392390010 392390050
ACRO392390010EA 38.2 EUR
ACRO392390010 ACRO392390050
4-Iodobiphenyl 97+%
4-Iodobiphenyl
Formula: C₁₂H₉I
MW: 280,11 g/mol
Boiling Pt: 320 °C
Melting Pt: 113 °C
Density: 1,584 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00019028
CAS Number: 1591-31-7
UN: 3077

Order Now

Specification Test Results

Appearance Off-white Crystalline powder
Infrared spectrum Conforms
Melting point 110 to 114 °C
HPLC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR