6-Chloronicotinonitrile 97%

Supplier: Thermo Fisher Scientific
Warning

398390050 398390250
ACRO398390050EA 38.3 EUR
ACRO398390050 ACRO398390250
6-Chloronicotinonitrile 97%
6-Chloronicotinonitrile
Formula: C₆H₃ClN₂
Boiling Pt: 107 °C (1 mmHg)
Melting Pt: 113…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00084941
CAS Number: 33252-28-7
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 116 to 120 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR