Azetidine hydrochloride 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Azetidine HCl
398730010
398730050
ACRO398730010EA
95.8
EUR
ACRO398730010
ACRO398730050
Azetidine hydrochloride 97%
Azetidine hydrochloride
Formula:
C₃H₈ClN MW: 93,56 g/mol Melting Pt: >300 °C Flash Pt: 152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191762 CAS Number: 36520-39-5 |
Specification Test Results
Appearance | White to brown Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...