4-Chloro-2-methoxybenzyl alcohol 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(4-Chloro-2-methoxyphenyl)methanol
398560010
398560050
ACRO398560010EA
37.9
EUR
ACRO398560010
ACRO398560050
4-Chloro-2-methoxybenzyl alcohol 97%
4-Chloro-2-methoxybenzyl alcohol
Formula:
C₈H₉ClO₂ MW: 172,61 g/mol Melting Pt: 47 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683547 CAS Number: 90296-27-8 |
Specification Test Results
Appearance | White to cream Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 47 to 51 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...