5-Bromoindole-2-carboxylic acid 98%

Supplier: Thermo Fisher Scientific
Warning

398650010 398650050 398652500
ACRO398650010EA 122 EUR
ACRO398650010 ACRO398650050 ACRO398652500
5-Bromoindole-2-carboxylic acid 98%
5-Bromoindole-2-carboxylic acid
Formula: C₉H₆BrNO₂
Storage Temperature: Freezer
MDL Number: MFCD00022705
CAS Number: 7254-19-5

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR