1-Boc-piperidine-3-carboxylic acid 98%

Supplier: Thermo Scientific
Warning

Synonyms: N-Boc-DL-nipecotic acid, 1-[(tert-butoxy)carbonyl]piperidine-3-carboxylic acid, 1-Boc-Nipecotic acid, Boc-Nip-OH, 1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid, N-BOC-3-Piperidinecarboxylic acid

399580050 399580250
ACRO399580050EA 47.4 EUR
ACRO399580050 ACRO399580250
1-Boc-piperidine-3-carboxylic acid 98%
1-Boc-piperidine-3-carboxylic acid
Formula: C₁₁H₁₉NO₄
MW: 229,3 g/mol
Melting Pt: 159…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00673775
CAS Number: 84358-12-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 159 to 162 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR