Isopropyl bromoacetate 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
isopropyl 2-bromoacetate, Acetic acid, bromo-, 1-methylethyl ester, Acetic acid, 2-bromo-, 1-methylethyl ester
412810250
412811000
ACRO412810250EA
121
EUR
ACRO412810250
ACRO412811000
Isopropyl bromoacetate 97%
Isopropyl bromoacetate
Formula:
C₅H₉BrO₂ MW: 181,029 g/mol Boiling Pt: 59…61 °C (10 mmHg) Density: 1,399 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009886 CAS Number: 29921-57-1 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4440 to 1.4460 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...