4-tert-Butylbenzaldehyde 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-(tert-Butyl)benzaldehyde

426520010 426520500 426522500
ACRO426520010EA 840 EUR
ACRO426520010 ACRO426520500 ACRO426522500
4-tert-Butylbenzaldehyde 95%
4-tert-Butylbenzaldehyde
Formula: C₁₁H₁₄O
Boiling Pt: 249 °C (760 mmHg)
Density: 0,97 g/cm³
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00035742
CAS Number: 939-97-9
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5270 to 1.5290 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR