Methyl undecanoate 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Undecanoic acid methyl ester, Undecylic acid methyl ester

426580050 426580250
ACRO426580050EA 57.2 EUR
ACRO426580050 ACRO426580250
Methyl undecanoate 97%
Methyl undecanoate
Formula: C₁₂H₂₄O₂
Boiling Pt: 246…248 °C (760 mmHg)
Density: 0,872 g/cm³
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00008957
CAS Number: 1731-86-8

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR