Indole-2-carbaldehyd 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Indole-2-carboxaldehyde, 1H-Indole-2-carbaldehyde
427510010
427510050
ACRO427510010EA
144
EUR
ACRO427510010
ACRO427510050
Indole-2-carbaldehyd 97%
Indole-2-carbaldehyd
Formula:
C₉H₇NO MW: 145,16 g/mol Melting Pt: 138…142 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03001425 CAS Number: 19005-93-7 |
Specification Test Results
Appearance | White to yellow to tan Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 133 to 143 °C |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...