Ethyl trans-β-methylcinnamate 97%
Supplier: Thermo Scientific
Synonyms:
Ethyl trans-beta-methylcinnamate, Ethyl (2E)-3-phenyl-2-butenoate
428000010
428000050
ACRO428000010EA
59
EUR
ACRO428000010
ACRO428000050
Ethyl trans-β-methylcinnamate 97%
Ethyl trans-β-methylcinnamate
Formula:
C₁₂H₁₄O₂ MW: 190,24 g/mol Boiling Pt: 142…144 °C (20hPa) Density: 1,042 g/cm³ Flash Pt: >113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053762 CAS Number: 1504-72-9 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
HPLC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...