N-Methyldicyclohexylamine 97%
Supplier: Thermo Scientific
|
Danger
|
428390010
428392500
ACRO428390010EA
274
EUR
ACRO428390010
ACRO428392500
N-Methyldicyclohexylamine 97%
N-Methyldicyclohexylamine
Formula:
C₁₃H₂₅N Boiling Pt: 265 °C (760 mmHg) Density: 0,921 g/cm³ Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014289 CAS Number: 7560-83-0 UN: 2735 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4890 to 1.4910 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...