Diazolidinyl urea 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1,3-Bis(hydroxymethyl)-5-[1,3-bis(hydroxymethyl)ureido]hydantoin, 1-(1,3-Bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl)-1,3-bis(hydroxymethyl)urea
428580250
428582500
ACRO428580250EA
55.7
EUR
ACRO428580250
ACRO428582500
Diazolidinyl urea 95%
Diazolidinyl urea
Formula:
C₈H₁₄N₄O₇ MW: 278,22 g/mol Melting Pt: 227…228 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03547942 CAS Number: 78491-02-8 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤3 % |
Solubility | (50 mg/ml in water) Clear colorless |
Elemental Analysis | 19.0 to 21.5 % N |
Hydroxyl value | 102 to 124 mg KOH/g |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...