2-Amino-6-chloropyridine 97%

Supplier: Thermo Scientific
Warning

Synonyms: 6-Chloro-2-pyridylamine, 6-Chloropyridin-2-amine

429500010 429500050
ACRO429500010EA 29.4 EUR
ACRO429500010 ACRO429500050
2-Amino-6-chloropyridine 97%
2-Amino-6-chloropyridine
Formula: C₅H₅ClN₂
MW: 128,56 g/mol
Melting Pt: 72…73,5 °C
Storage Temperature: Ambient
MDL Number: MFCD00234068
CAS Number: 45644-21-1

Order Now

Specification Test Results

Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Melting point 69 to 75 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR