1-Methyl-1H-indazol-5-amine 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-Amino-1-methyl-1H-indazole, 1-methyl-1H-indazole-5-amine, 1-METHYL-1H-INDAZOL-5-AMINE

429520010 429520050
ACRO429520010EA 97.8 EUR
ACRO429520010 ACRO429520050
1-Methyl-1H-indazol-5-amine 97%
1-Methyl-1H-indazol-5-amine
Formula: C₈H₉N₃
Melting Pt: 152…154 °C
Storage Temperature: Ambient
MDL Number: MFCD03305455
CAS Number: 50593-24-3

Order Now

Specification Test Results

Appearance Brown Crystalline powder
Infrared spectrum Conforms
Melting point 151 to 154 °C
HPLC ≥97.0 %
UV 1 cm cell vs water
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Acidity ≤0.0003 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR