Ethyl-2-nitrobenzoylacetate 97%

Supplier: Thermo Scientific

Synonyms: Benzenepropanoic acid,2-nitro-b-oxo-, ethyl ester, ethyl 3-(2-nitrophenyl)-3-oxopropanoate, 2-Nitrobenzoylacetic acid ethyl ester

430040010 430040050
ACRO430040010EA 103 EUR
ACRO430040010 ACRO430040050
Ethyl-2-nitrobenzoylacetate 97%
Ethyl-2-nitrobenzoylacetate
Formula: C₁₁H₁₁NO₅
MW: 237,211 g/mol
Melting Pt: 35…36 °C
Storage Temperature: Ambient
MDL Number: MFCD03424821
CAS Number: 52119-39-8

Order Now

Specification Test Results

Appearance Clear orange to brown Liquid or low melting solid
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR