N,N-Dimethylacrylamide 99% stabilised

Supplier: Thermo Fisher Scientific
Danger


Stabilisation: Stabilised with 4-methoxyphenol (MEHQ) 500 ppm

432021000 432025000
ACRO432021000EA 85.7 EUR
ACRO432021000 ACRO432025000
N,N-Dimethylacrylamide 99% stabilised
N,N-Dimethylacrylamide
Formula: C₅H₉NO
Boiling Pt: 80 °C (20 mmHg)
Density: 0,962 g/cm³
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008626
CAS Number: 2680-03-7
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4730 to 1.4750 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer Approx. 500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR