6-Bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one 97%

Supplier: Thermo Fisher Scientific
Warning

432710010 432710050
ACRO432710010EA 203 EUR
ACRO432710010 ACRO432710050
6-Bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one 97%
6-Bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one
Formula: C₈H₆BrNO₂
Melting Pt: 149…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00694789
CAS Number: 67927-44-0

Order Now

Specification Test Results

Appearance Off-white Powder
Infrared spectrum Conforms
Melting point 147 to 151 °C
GC ≥96.0 %
Specific optical rotation +63° to +67° (20 °C, 589 nm) (c=4, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR